Glacier Ice Buckets, Electron Microscopy Sciences | Avantor
Glacier Ice Buckets, Electron Microscopy Sciences
10.625"
Catalog # 102096-374
Glacier Ice Buckets, Electron Microscopy Sciences
Catalog # 102096-374
Supplier:  
Glacier Ice Buckets, Electron Microscopy Sciences
Glacier Ice Buckets, Electron Microscopy Sciences
10.625"
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
VWR BUCKET/PAN ICE W/LID STCK PU GR 4.5L Catalog #10146-192
Le produit 10146-192 est similaire à 102096-374 mais peut avoir un fabricant et/ou un emballage différents.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Excellent Insulation: Made of the same triple density expanded polystyrene as our popular Ice Pans, preserve ice 30% longer.
  • Won't Sweat - Thick insulating walls will not sweat
  • Easy Grip - Robust handles make buckets easy to carry
  • Includes Lid - Use the supplied lid to maximize cooling
  • Heavy Duty - Solidly constructed to be unbreakable under normal laboratory conditions
  • Chemically Resistant - Resistant to mild chemicals* as well as ice-alcohol slurry, dry ice and liquid nitrogen
(*Not recommended for long-term exposure to harsh chemicals or corrosives.)
Spécifications
  • Top Diameter:
    10.625"