Mini-Cooling Chambers, Electron Microscopy Sciences | Avantor
Mini-Cooling Chambers, Electron Microscopy Sciences
Mini Cooling Chamber 30 Holes
Catalog # 102096-366
Fournisseur #:  68600-30
Mini-Cooling Chambers, Electron Microscopy Sciences
Catalog # 102096-366
Supplier:  
Mini-Cooling Chambers, Electron Microscopy Sciences
Mini-Cooling Chambers, Electron Microscopy Sciences
Mini Cooling Chamber 30 Holes
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ideal for ice pans and ice buckets for use when only a small number of microtubes are required.
Each Mini-Chamber will fit in our 1L Glacierbrand™ Ice Pan (6.13 x 6.13 x 3.61"). Minimize Ice - Less Ice is needed to cool these chambers. Less ice means less mess. Mini-Chambers are anodized aluminum and utilize our "Surround-A-Tube" cooling technology.
Spécifications
  • Désignation:
    Mini Cooling Chamber 30 Holes