Glow Writer, Electron Microscopy Sciences | Avantor
Glow Writer, Electron Microscopy Sciences
Glow Writer Pen
Catalog # 102096-326
Fournisseur #:  68551-01
Glow Writer, Electron Microscopy Sciences
Catalog # 102096-326
Glow Writer, Electron Microscopy Sciences
Glow Writer, Electron Microscopy Sciences
Glow Writer Pen
Price
$198.05
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Fine-point phosphorescent marking pen for laboratory autography.
Glow Writer is a phosphorescent pen for marking chemiluminescent and isotope-labeled substrates for X-ray film autography. The unique marker delivers a phosphor-containing ink through a fine point porous nib. Before exposure to X-ray film, the phosphorescent writing is excited by room light. The pen utilizes a phosphor ink having an optimum concentration of phosphor and a relatively short afterglow half-life (approximately 2-3 minutes) has been selected so that upon exposure to X-ray film, autographic exposures of almost any duration will be easily readable on the film. The fine point nib is easily replaceable if it dries out.
Spécifications
  • Désignation:
    Glow Writer Pen