Dual Micrometer Calibration Slide, Electron Microscopy Sciences
EMS has introduced two new calibration slides that have the benefit of dual imperial/metric scales.
- New Dual-Scale Calibration Slides
- 2" Imperial (English) and 50mm Metric Scales on a Single Slide
- Positive and Negative Versions
- Unique Serial Number for Traceability
- Available with Internationally Traceable Certificates of Calibration
The PS52P is for transmitted light applications and has a bright chrome positive image. The PS52N has a negative pattern, formed in low reflective chrome for incident light applications to give excellent contrast. Both are ideal for calibrating optical products with a large field of view, such as stereo microscopes or imaging systems. Standard and Crossed Patterns – S1, S2, S4, S5, S8, S11, S12, S16, S20, S21, S22, S48.
- Désignation:Dual micrometer calibration slides PS52N with ukas certificate