Glass Rods, Electron Microscopy Sciences | Avantor
Glass Rods, Electron Microscopy Sciences
Glass Rods, 250 mm
Catalog # 102095-964
Fournisseur #:  66206-25
Glass Rods, Electron Microscopy Sciences
Catalog # 102095-964
Glass Rods, Electron Microscopy Sciences
Glass Rods, Electron Microscopy Sciences
Glass Rods, 250 mm
Price
$20.89
Log in pour la disponibilité
Each Pack of 12
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Useful tool for all laboratory stirring and mixing
  • 200 mm and 250 mm lengths available
  • Policeman is rubber cover helps help stir at the bottom of container
Useful tool for all laboratories needs to stir/mix substances
Policeman is rubber cover to fit 5 mm rods above. The sealed end is cut at an angle to help stir at the bottom of container.
Glass rods are 5 mm diameter, and come in three lengths.
Spécifications
  • Désignation:
    Glass Rods
  • Longueur:
    250 mm
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Rubber Policeman
    Rubber Policeman
    102095-968
    Log in pour la disponibilité
    Each - $57.05