Single and Multi Wafer Containers, Electron Microscopy Sciences
Single and Multi Wafer Containers, Electron Microscopy Sciences
Wafer Container Multi, —, 100 mm (4")
Catalog # 102095-804
Supplier #:  64918-4
Single and Multi Wafer Containers, Electron Microscopy Sciences
Catalog # 102095-804
Single and Multi Wafer Containers, Electron Microscopy Sciences
Single and Multi Wafer Containers, Electron Microscopy Sciences
Wafer Container Multi, —, 100 mm (4")
Price
$398.07
Log in for availability
Each Pack of 3
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These containers are for storage and shipping of a variety of sizes of wafers (Any material) from 1 to 6" in diameter. They prevent damage and contamination to the wafer. Features a conical base to support the wafers on their edges. Wafers are held in place by a spring.
  • Conical base to support the wafers on their edges
  • Wafers are held in place by the spring
The multi-wafer containers are made from polypropylene and have a storage capacity of 25 wafers maximum. They come complete with the base and cover. The single containers will accommodate a variety of sizes from 4 to 6" by simply adjusting the silicone supports in the box. These containers are made from anti-electrostatic polycarbonate and are available in transparent or black.
 : The set includes the container, the cover and the spring.
Specifications
  • Désignation:
    Wafer Container Multi
  • Taille:
  • For Use With:
    100 mm (4")