Single and Multi Wafer Containers, Electron Microscopy Sciences
Wafer Container Multi, —, 75 mm (3")
Catalog # 102095-802
Supplier: Electron Microscopy Sciences
: 64918-3
Single and Multi Wafer Containers, Electron Microscopy Sciences
Catalog # 102095-802
Supplier: Electron Microscopy Sciences
Jump to:
Product Details & Documents
These containers are for storage and shipping of a variety of sizes of wafers (Any material) from 1 to 6" in diameter. They prevent damage and contamination to the wafer. Features a conical base to support the wafers on their edges. Wafers are held in place by a spring.
- Conical base to support the wafers on their edges
- Wafers are held in place by the spring
The multi-wafer containers are made from polypropylene and have a storage capacity of 25 wafers maximum. They come complete with the base and cover. The single containers will accommodate a variety of sizes from 4 to 6" by simply adjusting the silicone supports in the box. These containers are made from anti-electrostatic polycarbonate and are available in transparent or black.
: The set includes the container, the cover and the spring.
Spécifications
- Désignation:Wafer Container Multi
- Taille:—
- For Use With:75 mm (3")