GEL-BOX (AD Carrier Boxes), Electron Microscopy Sciences
GEL-BOX (AD Carrier Boxes), Electron Microscopy Sciences
Conductive BD Tray in Antistatic Hinged Box, 2 x 2"
Catalog # 102095-742
Supplier #:  64900-10
GEL-BOX (AD Carrier Boxes), Electron Microscopy Sciences
Catalog # 102095-742
Supplier:  
GEL-BOX (AD Carrier Boxes), Electron Microscopy Sciences
GEL-BOX (AD Carrier Boxes), Electron Microscopy Sciences
Conductive BD Tray in Antistatic Hinged Box, 2 x 2"
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Gel-Slide (coated glass slides) is designed for higher temperature applications such as bonding and reflow where a plastic tray will not work.
Same as the Gel-Box, the Gel-Tray and Gel-Slide products are intended for manual applications in which the devices are loaded and unloaded using tweezers or by fingers. The Gel-Pak BD and CD series of pocketless trays are both configured as a 2" carrier stored in a protective plastic hinged box. The Gel coating is applied directly to the surface of the plastic tray (BD Series) or glass slide (CD Series). These products offer the same functionality as the Gel-Box, however the plastic tray or glass slide can be easily removed from the box allowing for convenient handling when loading or unloading fixtures are being used.
The Gel-Slide (coated glass slides) is designed for higher temperature applications such as bonding and reflow where a plastic tray will not work. The CD slide has a temperature capability up to 220 °C. The CD glass slides are also ideal for backside inspection due to their optical clarity. Both the Gel-Tray and Gel-Slide products are available in the standard retention levels (X0, X4, X5, and X8) to accommodate variations in device size, weight, and surface roughness. The protective 2" hinged boxes are available in transparent styrene (T), conductive black (C) and antistatic (AS) materials. The boxes, trays, and slides may be customized with a print pattern (grid) or company logo. Standard available Prints can be viewed at Gel-Tray/Gel-Slide Print options.
Specifications
  • Désignation:
    Conductive BD Tray in Antistatic Hinged Box
  • Taille:
    2 x 2"