Nunc™ 384 Well® Optical Bottom Plates – Polymer Base, Electron Microscopy Sciences
The NUNC™ Coverglass Optical Bottom 96- and 384- Well Plates have an optically clear polystyrene plate bottom
This clear base permits microscopic viewing and reading of well The plate’s upper structure is offered in black (for fluorometric detection).
- Color:White
- Désignation:Non-Sterile with lid
- Dimensions:128 × 86 mm
- Surface Treatment:Collagen I
- Working Well Volume:10 - 100 µl