Micro Tissue Grinder Kit, Electron Microscopy Sciences
Micro Tissue Grinder Kit, Electron Microscopy Sciences
Tissue Grinder, 0.2 ml
Catalog # 102095-700
Supplier #:  64799-02
Micro Tissue Grinder Kit, Electron Microscopy Sciences
Catalog # 102095-700
Micro Tissue Grinder Kit, Electron Microscopy Sciences
Micro Tissue Grinder Kit, Electron Microscopy Sciences
Tissue Grinder, 0.2 ml
Price
$423.99
Log in for availability
Each Pack of 2
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This complete selection of Wheaton Micro Tissue Grinders is conveniently packed in a lightweight protective carrying case. The 0.5 ml micro tissue grinder with screw cap can be used for tissue grinding as well as for additional procedures.
  • Complete kit includes 7 micro tissue grinders
  • Made from Wheaton 33 low extractable borosilicate glass
  • Replacement parts available separately
  • Autoclavable
This particular tissue grinder mortar can be safely joined with another threaded glass component if needed, using the Wheaton Connection®. The Kit is packed in high-density polyethylene case with foam inserts.
Specifications
  • Désignation:
    Tissue Grinder
  • Taille:
    0.2 ml