Potter-Elvehjem Tissue Grinder, Electron Microscopy Sciences
Potter-Elvehjem Tissue Grinder, Electron Microscopy Sciences
Pestle only Tissue Grinders, 15 ml
Catalog # 102095-582
Supplier #:  64792-83
Potter-Elvehjem Tissue Grinder, Electron Microscopy Sciences
Catalog # 102095-582
Supplier:  
Potter-Elvehjem Tissue Grinder, Electron Microscopy Sciences
Potter-Elvehjem Tissue Grinder, Electron Microscopy Sciences
Pestle only Tissue Grinders, 15 ml
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These grinder are precisely constructed with completely interchangeable parts to allow easier cleaning or replacement of damaged mortars. They are designed for use in preparation of tissue homogenates and other similar applications.
  • Ideal for tissue homogenates
  • Use with power homogenization
  • Parts interchangeable
  • Teflon® pestle
  • Autoclavable
The Teflon pestle is for soft tissues such as brain or liver. The stainless steel rod is 0.25" (6.3mm) diameter. The grinding chamber clearance is 0.004" to 0.006’ (0.1 to 0.15mm). This unit is autoclavable and can be used for power homogenization as with overhead stirrer. However, we strongly recommend the use of safety-coated tissue grinder (64793-series) for added safety. This unit is manufactured from Wheaton 33 low extractable borosilicate glass that conforms to USP Type 1 and ASTM E-438 Type 1, Class A requirements.
Spécifications
  • Désignation:
    Pestle only Tissue Grinders
  • Taille:
    15 ml