Dounce Tissue Grinder, Electron Microscopy Sciences | Avantor
Dounce Tissue Grinder, Electron Microscopy Sciences
Dounce Tissue Grinder, 7 ml
Catalog # 102095-540
Fournisseur #:  64790-07
Dounce Tissue Grinder, Electron Microscopy Sciences
Catalog # 102095-540
Dounce Tissue Grinder, Electron Microscopy Sciences
Dounce Tissue Grinder, Electron Microscopy Sciences
Dounce Tissue Grinder, 7 ml
Price
$1,050.26
Log in pour la disponibilité
Each Pack of 2
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This grinder designed to retain a high percentage of cell nuclei and mitochondria in soft tissues from cell cultures. When use, the pestle ball is encircled in liquid, which is avoid heat buildup by reducing friction.
  • Ideal for soft tissue
  • Provide with loose and tight pestle
  • Wheaton 33 low extractable borosilicate glass
  • Autoclavable
This grinder is ideal for enzyme studies. The initial grinding is performed using the “loose” pestle. The grinding process is completed using the “tight” pestle.. This grinder is used by moving the pestle up and down, and work best with cell suspensions or very soft tissues. The mortar has a large reservoir and pouring lip and is supplied with “loose” and “tight” pestles. Tight Pestle “A” Specification: 0.0010 – 0.0030” (0.025 – 0.076 mm) Loose Pestle “B” Specification: 0.0035 – 0.0055” (0.089 – 0.14 mm)
Spécifications
  • Désignation:
    Dounce Tissue Grinder
  • Taille:
    7 ml