Safety Goggles, Electron Microscopy Sciences
Perforated frame small size, 5³/₄×2¹/₂"
Catalog # 102095-490
Supplier #: 64477-01
Jump to:
Discontinued
Product Details & Documents
These safety googles are recommended for use in cryo preparation.
- Avoid chemical contact
- Scratch resistant lenses
Specifications
- Désignation:Perforated frame small size, 5³/₄×2¹/₂"