Lab Drawer Organizers, Electron Microscopy Sciences | Avantor
Lab Drawer Organizers, Electron Microscopy Sciences
Small Parts Tray
Catalog # 102095-158
Fournisseur #:  64370-59
Lab Drawer Organizers, Electron Microscopy Sciences
Catalog # 102095-158
Lab Drawer Organizers, Electron Microscopy Sciences
Lab Drawer Organizers, Electron Microscopy Sciences
Small Parts Tray
Price
$189.02
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
We offer a variety of trays for your drawers or to fill in 6-drawer and 7-shelf carts.
  • Trays can store and sort supplies of all sizes and shapes
Trays are made of ABS, color white, and chemical resistant.
Spécifications
  • Désignation:
    Small Parts Tray