Extra Deep Trays, Electron Microscopy Sciences | Avantor
Extra Deep Trays, Electron Microscopy Sciences
Catalog # 102095-140
Extra Deep Trays, Electron Microscopy Sciences
Catalog # 102095-140
Supplier:  
Extra Deep Trays, Electron Microscopy Sciences
Extra Deep Trays, Electron Microscopy Sciences
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These deep trays with dividers, can organize your larger and bulkier supplies.
  • Measures L×W: 17.5×19.5"
  • With heights of either 4, 6, or 8"
They are constructed of bright white plastic with smooth surface.
Available with three depths: 4, 6, and 8 inches, and with or without hinged clear lid.