Glass Sample Bottles, With Plastic Snap Cap, Electron Microscopy Sciences | Avantor
Glass Sample Bottles, With Plastic Snap Cap, Electron Microscopy Sciences
Replacement Snap Cap, 30 mm
Catalog # 102095-026
Fournisseur #:  64257-30
Glass Sample Bottles, With Plastic Snap Cap, Electron Microscopy Sciences
Catalog # 102095-026
Glass Sample Bottles, With Plastic Snap Cap, Electron Microscopy Sciences
Glass Sample Bottles, With Plastic Snap Cap, Electron Microscopy Sciences
Replacement Snap Cap, 30 mm
Price
$138.47
Log in pour la disponibilité
Each Pack of 200
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For collecting and exhibiting a wide variety of specimens in the lab or field. Caps snap over a special sealing ring to form an effective barrier to moisture, making these bottles especially suitable for hydroscopic materials.
Spécifications
  • Désignation:
    Replacement Snap Cap
  • Taille:
    30 mm