Tri-Seal™ container, Electron Microscopy Sciences
Product Details & Documents
Tri-Seal™ Specimen Container is a patented, leakproof, airtight, touch-top specimen container for the pathology lab.
Vials have permanent hinge and triple seal design. These are molded from 100% High-Density, translucent polypropylene and are available in a variety of sizes from 13 ml to 300 ml.
Spécifications
- Capacité:240 ml
- Dimensions:75 × 75 mm