Tri-Seal™ container, Electron Microscopy Sciences
13 ml, 15 × 95 mm
  102094-990
 :  64235-13
Tri-Seal™ container, Electron Microscopy Sciences
  102094-990
 :  
Tri-Seal™ container, Electron Microscopy Sciences
Tri-Seal™ container, Electron Microscopy Sciences
13 ml, 15 × 95 mm
Tri-Seal™ Specimen Container is a patented, leakproof, airtight, touch-top specimen container for the pathology lab.
Vials have permanent hinge and triple seal design. These are molded from 100% High-Density, translucent polypropylene and are available in a variety of sizes from 13 ml to 300 ml.
  • Capacité:
    13 ml
  • Dimensions:
    15 × 95 mm