Tri-Seal™ container, Electron Microscopy Sciences | Avantor
Tri-Seal™ container, Electron Microscopy Sciences
15 ml, 21 × 60 mm
Catalog # 102094-976
Fournisseur #:  64234-15
Tri-Seal™ container, Electron Microscopy Sciences
Catalog # 102094-976
Tri-Seal™ container, Electron Microscopy Sciences
Tri-Seal™ container, Electron Microscopy Sciences
15 ml, 21 × 60 mm
Price
$1,396.96
Log in pour la disponibilité
Each Pack of 500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Tri-Seal™ Specimen Container is a patented, leakproof, airtight, touch-top specimen container for the pathology lab.
Vials have permanent hinge and triple seal design. These are molded from 100% High-Density, translucent polypropylene and are available in a variety of sizes from 13 ml to 300 ml.
Spécifications
  • Capacité:
    15 ml
  • Dimensions:
    21 × 60 mm