C-Chip Disposable Hemacytometer, Electron Microscopy Sciences | Avantor
C-Chip Disposable Hemacytometer, Electron Microscopy Sciences
Cellometer Disposable Counting Chamber, Plastic Base
Catalog # 102094-790
Fournisseur #:  63561-25
C-Chip Disposable Hemacytometer, Electron Microscopy Sciences
Catalog # 102094-790
C-Chip Disposable Hemacytometer, Electron Microscopy Sciences
C-Chip Disposable Hemacytometer, Electron Microscopy Sciences
Cellometer Disposable Counting Chamber, Plastic Base
Price
$477.09
Log in pour la disponibilité
Box of 25 Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
With a pipetor, 15 to 20 microlilter cell-containing sample is introduced into the counting chamber through one of the ports
These Disposable Cell Counters are an enclosed chamber with two ports for sample introduction. The chamber contains precisely spaced lines in a grid pattern. There are two counting chambers per slide for replicates The counting grid pattern is the Improved Neubauer, as in a common Hemacytometer. The consistent pattern design allows the standard cell counting procedure to be followed.
With a pipetor, 15 to 20 microlilter cell-containing sample is introduced into the counting chamber through one of the ports. Liquid spreads inside the chamber, replaces air, which escapes through the opposing port. The cell counting chamber is placed under a phase contrast optical microscope with a typical magnification of 100. The microscope is focused both onto the grid pattern and the cell particles. Standard protocols are followed for cell counting.
Liquid spreads inside the chamber, replaces air, which escapes through the opposing port. The cell counting chamber is placed under a phase contrast optical microscope with a typical magnification of 100. The microscope is focused both onto the grid pattern and the cell particles. Standard protocols are followed for cell counting.
Spécifications
  • Désignation:
    Cellometer Disposable Counting Chamber, Plastic Base