Double Neubauer Ruled Hemacytometer, Electron Microscopy Sciences | Avantor
Double Neubauer Ruled Hemacytometer, Electron Microscopy Sciences
Double Neubauer Chamber with 2 Coverglass (3100)
Catalog # 102094-780
Fournisseur #:  63510-11
Double Neubauer Ruled Hemacytometer, Electron Microscopy Sciences
Catalog # 102094-780
Double Neubauer Ruled Hemacytometer, Electron Microscopy Sciences
Double Neubauer Ruled Hemacytometer, Electron Microscopy Sciences
Double Neubauer Chamber with 2 Coverglass (3100)
Price
$1,279.05
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The complete set includes one counting chamber, two 0.5 mm cover glasses, one red cell pipette, one white blood cell pipette, tubing, mouthpieces and carrying case.
An “H” shape moat which forms 2 counting areas are metal plated and double Neubauer ruled providing sharp bright line contrast for consistent cell counts. A “V” cut on each area is used to control the possibility of overflow. Triple dividing lines clearly show cells within the counting area. The ruled surface is 0.1 mm below the cover glass; liquid capacity is 0.1 mm3/mm2 and 0.00025 mm3/grid square.
The complete set includes one counting chamber, two 0.5 mm cover glasses, one red cell pipette, one white blood cell pipette, tubing, mouthpieces and carrying case.
Spécifications
  • Désignation:
    Double Neubauer Chamber with 2 Coverglass (3100)