Flexwell™ Incubation Chambers, Electron Microscopy Sciences
Product Details & Documents
FlexWell™ removable incubation chambers for microarrays using ONCYTE® slides.
- Silicone FlexWell™ forms well on slides using a clean release adhesive to isolate up to 16 specimens per slide
- Slides and FlexWell™ fit into a reusable slide tray organizing wells into the footprint of a SBS compliant microtiter plate
- Gasket may be sealed for mixing and to prevent evaporation
- Available in eight-well and 16-well formats
Spécifications
- Profondeur:—
- Ø ext.:75.9 x 127.6 mm
- Désignation:4 Slide Tray