ProPlate™ Microtiter Plate, Electron Microscopy Sciences
ProPlate™ Microtiter Plate, Electron Microscopy Sciences
ONCYTE AVID® 96 NC Round Pad, 6 mm dia, Plastic Substrate
Catalog # 102094-742
Supplier #:  63485-22
ProPlate™ Microtiter Plate, Electron Microscopy Sciences
Catalog # 102094-742
Supplier:  
ProPlate™ Microtiter Plate, Electron Microscopy Sciences
ProPlate™ Microtiter Plate, Electron Microscopy Sciences
ONCYTE AVID® 96 NC Round Pad, 6 mm dia, Plastic Substrate
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Patented  ProPlate™ technology for high throughput robotic processing of microarrays
The proprietary adhesive forms a water-tight seal between a microarray substrate and the bottomless microtiter plate. Grace Bio Labs offers plate with SBS standard spacing in both 96-well (shown) and 384-well formats.
Key product features include:
Guaranteed flat SBS compliant microtiter plate format which enables robotic processing of assays
Water-tight seal between superstructure and substrate
Compatible with glass or plastic substrates
Available in 96 round or square or 384 square well format
Available as a complete microtiter plate with microcellulose film coated substrate, uncoated substrate or as an adhesive superstructure alone
Plates are available with SBS standard spacing in both 96-well and 384-well formats
Grace BioLabs developed bottomless SBS compliant microtiter plate superstructures for use with robotic processors. TheSuperstructures come in a permanently affixed well format – the ProPlate MP™, or a removable well format – the FlexPlate MP™. The ProPlate MP™ SBS compliant microtiter plate features a SecureSeal™ Plus adhesive bonding system applied to a bottomless 96-round well, black, polystyrene microtiter plate. Simply peel off the protective polyliner and apply large format array for automated processing.
Specifications
  • Désignation:
    ONCYTE AVID® 96 NC Round Pad, 6 mm dia, Plastic Substrate