ProPlate™ Microtiter Plate, Electron Microscopy Sciences
Product Details & Documents
Patented ProPlate™ technology for high throughput robotic processing of microarrays
The proprietary adhesive forms a water-tight seal between a microarray substrate and the bottomless microtiter plate. Grace Bio Labs offers plate with SBS standard spacing in both 96-well (shown) and 384-well formats.
Key product features include:
Guaranteed flat SBS compliant microtiter plate format which enables robotic processing of assays
Water-tight seal between superstructure and substrate
Compatible with glass or plastic substrates
Available in 96 round or square or 384 square well format
Available as a complete microtiter plate with microcellulose film coated substrate, uncoated substrate or as an adhesive superstructure alone
Plates are available with SBS standard spacing in both 96-well and 384-well formats
Grace BioLabs developed bottomless SBS compliant microtiter plate superstructures for use with robotic processors. TheSuperstructures come in a permanently affixed well format – the ProPlate MP™, or a removable well format – the FlexPlate MP™. The ProPlate MP™ SBS compliant microtiter plate features a SecureSeal™ Plus adhesive bonding system applied to a bottomless 96-round well, black, polystyrene microtiter plate. Simply peel off the protective polyliner and apply large format array for automated processing.
Spécifications
- Désignation:ONCYTE AVID® 96 NC Round Pad, Glass Substrate