ProPlate™ MicroArray System, Electron Microscopy Sciences | Avantor
ProPlate™ MicroArray System, Electron Microscopy Sciences
3 Well Module, 18x20 mm Well, Stainless Steel Clip
Catalog # 102094-690
Fournisseur #:  63484-43
ProPlate™ MicroArray System, Electron Microscopy Sciences
Catalog # 102094-690
Supplier:  
ProPlate™ MicroArray System, Electron Microscopy Sciences
ProPlate™ MicroArray System, Electron Microscopy Sciences
3 Well Module, 18x20 mm Well, Stainless Steel Clip
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
ProPlate™ MicroArray System is an integration of microplate and biochip technologies
Each ProPlate Tray Set includes: 4 Slide modules, 8 Snap clips, 1 Tray and Cover, 10 seal strips and one applicator. Tray conforms to SBS standards for footprint (81.7mm width x 123.7mm length) and well spacing. System available in Chip Chip™ compatible formats.
ProPlate™ integrates microscope slide based array technology with automated microtiter plate processing. Up to 64 individual arrays can be assayed per slide. Four slides fit into a tray, producing a modular plate with a standard microtiter plate footprint and well spacing allowing automated robotic processing up to 256 arrays.
Spécifications
  • Désignation:
    3 Well Module, 18x20 mm Well, Stainless Steel Clip