Forceps Jar, Electron Microscopy Sciences | Avantor
Forceps Jar, Electron Microscopy Sciences
Forceps jar, 8", 4"
Catalog # 102094-604
Fournisseur #:  63313-07
Forceps Jar, Electron Microscopy Sciences
Catalog # 102094-604
Supplier:  
Forceps Jar, Electron Microscopy Sciences
Forceps Jar, Electron Microscopy Sciences
Forceps jar, 8", 4"
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Convenient container for keeping forceps handy.
  • 3 and 4 inch diameters available
  • 3, 4, 5, and 8 inch heights
Made of stainless steel with wide base and polished finished.
Spécifications
  • Ø:
    4"
  • Désignation:
    Forceps jar
  • Hauteur:
    8"