Ice Pan, Electron Microscopy Sciences | Avantor
Ice Pan, Electron Microscopy Sciences
6.13"
Catalog # 102094-590
Ice Pan, Electron Microscopy Sciences
Catalog # 102094-590
Supplier:  
Ice Pan, Electron Microscopy Sciences
Ice Pan, Electron Microscopy Sciences
6.13"
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
VWR BUCKET/PAN ICE W/LID STCK BK SM 1L Catalog #10146-226
Le produit 10146-226 est similaire à 102094-590 mais le fabricant et les spécifications peuvent varier.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
*Not recommended for long-term exposure to harsh chemicals or corrosives.
Half the price of competitor's ice pans. Thick pan walls will not sweat or leave puddles of condensation on the benchtop. Made of triple density expanded polystyrene creating a durable container that is virtually unbreakablePreserves ice up to 30% longer than conventional ice pans. Designed with an inner edge enabling the pans to stack on top of one another for easy storage and to minimize bench-top space. Ice pans are extremely lightweight (1.1, 4.7. and 11.4 oz.) and easy to handle. They feature a molded lip around the edge for carefree moving and carrying. Resistant* to mild chemicals as well as ice-alcohol slurry, dry ice and liquid nitrogen.
*Not recommended for long-term exposure to harsh chemicals or corrosives.
Spécifications
  • Profondeur:
    6.13"