Floatation Bath & Flotation Workstations, Electron Microscopy Sciences
This Flotation Workstation helps to make your tissue preparation easier, safer and more efficient.
- Magnetic non-contact switch function for years flawlessly
- Microprocessor-controlled heating, ambient to 65°C
- Flat membrane switches for safety and simplicity
- Fluorescent white lighting positioned for improve tissue viewing glare-free
- Heat-resistant plastic housing maintains cosmetic appearance
- Removable glass dish for easy cleaning
- 2 x 6" or 2 x 8" Convection Slide dryer & 2 x 2" HISTO Orientator™ for eliminating wrinkles
The unit comes with conveniently removable glass dish, scratch and chemical resistant plastic housing, microprocessor controlled electronics and a high contrast background for optimum specimen viewing combined with the HISTO Orientator™ for flattening sections and a Slide Dryer, all in one compact unit. The systems are now available in Low Profile 8 x 8 x 2 ¼" deep dish versions as well as for those labs wanting to float larger ribbons, 8 x 11 x 2".
For those flotation bath applications not requiring the add convenience of Slide Dryer and HISTO Orientator™, the basic Flotation Bath provides all the functional need of the Flotation Workstation at the affordable price. The baths again come in two sizes , 8 x 8 x 2 1/4" or 8 x 11 x 2".
- Désignation:Slide Drying Rack, Front mounted
- Taille:8 x 11" Dish