Floatation Bath & Flotation Workstations, Electron Microscopy Sciences | Avantor
Floatation Bath & Flotation Workstations, Electron Microscopy Sciences
Workstation 2 x 6" Slide Dryer, 120 VAC, 8 x 8 x 2¼" Dish
Catalog # 102094-526
Fournisseur #:  63214-10
Floatation Bath & Flotation Workstations, Electron Microscopy Sciences
Catalog # 102094-526
Supplier:  
Floatation Bath & Flotation Workstations, Electron Microscopy Sciences
Floatation Bath & Flotation Workstations, Electron Microscopy Sciences
Workstation 2 x 6" Slide Dryer, 120 VAC, 8 x 8 x 2¼" Dish
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
FLOTATION WORK STATION 8X8X2 Catalog #89207-128
Le produit 89207-128 est similaire à 102094-526 mais le fabricant et les spécifications peuvent varier.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This Flotation Workstation helps to make your tissue preparation easier, safer and more efficient.
  • Magnetic non-contact switch function for years flawlessly
  • Microprocessor-controlled heating, ambient to 65°C
  • Flat membrane switches for safety and simplicity
  • Fluorescent white lighting positioned for improve tissue viewing glare-free
  • Heat-resistant plastic housing maintains cosmetic appearance
  • Removable glass dish for easy cleaning
  • 2 x 6" or 2 x 8" Convection Slide dryer & 2 x 2" HISTO Orientator™ for eliminating wrinkles
The unit comes with conveniently removable glass dish, scratch and chemical resistant plastic housing, microprocessor controlled electronics and a high contrast background for optimum specimen viewing combined with the HISTO Orientator™ for flattening sections and a Slide Dryer, all in one compact unit. The systems are now available in Low Profile 8 x 8 x 2 ¼" deep dish versions as well as for those labs wanting to float larger ribbons, 8 x 11 x 2".
For those flotation bath applications not requiring the add convenience of Slide Dryer and HISTO Orientator™, the basic Flotation Bath provides all the functional need of the Flotation Workstation at the affordable price. The baths again come in two sizes , 8 x 8 x 2 1/4" or 8 x 11 x 2".
Spécifications
  • Désignation:
    Workstation 2 x 6" Slide Dryer, 120 VAC
  • Taille:
    8 x 8 x 2¼" Dish