CoolSafe Cool Bricks, Electron Microscopy Sciences | Avantor
CoolSafe Cool Bricks, Electron Microscopy Sciences
1°C Cool Brick
Catalog # 102093-846
Fournisseur #:  61952-30
CoolSafe Cool Bricks, Electron Microscopy Sciences
Catalog # 102093-846
Supplier:  
CoolSafe Cool Bricks, Electron Microscopy Sciences
CoolSafe Cool Bricks, Electron Microscopy Sciences
1°C Cool Brick
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Each Cool Brick allows up to 10 hours of benchtop temperature stability.
Two temperature zones are available: 1°C and -10°C.
Spécifications
  • Désignation:
    1°C Cool Brick