CoolSafe Cool Bricks, Electron Microscopy Sciences
Product Details & Documents
Each Cool Brick allows up to 10 hours of benchtop temperature stability.
Two temperature zones are available: 1°C and -10°C.
Specifications
- Désignation:-10°C Cool Brick