Microtube Rack, 4-Way, Electron Microscopy Sciences
Create as large a rack as you need by linking together multiple racks. Heavyduty polypropylene racks feature a unique system of tabs and slots, that facilitate easy connection and sturdy fit. Autoclavable. Assorted colors or natural. Measurements: 174 x 95 x 52 mm.
- Désignation:4-Way Microtube Rack Natural