96-Well Reversible Rack, Electron Microscopy Sciences
96-Well Reversible Rack, Electron Microscopy Sciences
Yellow
  102093-658
Supplier #:  61602-Y
96-Well Reversible Rack, Electron Microscopy Sciences
  102093-658
 :  
96-Well Reversible Rack, Electron Microscopy Sciences
96-Well Reversible Rack, Electron Microscopy Sciences
Yellow
RACK REVERSIBLE W/COVER 96-PLACE YELLOW  76169-950
Product 76169-950 is similar to 102093-658 but manufacturer and specifications may vary.
96 PLAC MIC RAC POLY YEL PK5  21150-240
Product 21150-240 is similar to 102093-658 but manufacturer and specifications may vary.
Polypropylene rack has 96 wells to accommodate 0.5 mL micro centrifuge tubes on one side, or 96 wells for 1.5 to 2.0 mL micro centrifuge tubes on the other side. Arranged in standard 8 x 12 array. Handy lid snaps into place in the tabs on each end of rack. Grid lines are molded on the side of the rack that accommodates the smaller tubes making it easier to see the wells. Raised rings are molded on the opposite side of the rack making it easier to replace the 1.5 to 2.0 mL tubes from those wells. Rack measures 9.75 x 4.75 x 1.875".
  • Color:
    Yellow