81-Well Storage Rack, Electron Microscopy Sciences
Product Details & Documents
Durable polypropylene rack fits in standard 5.25” square freezer modules and holds 1.5 to 2.0 ml microtubes and cryovials.
- Molded grid lines on the lid
- Imprinted coordinates on the bottom of base
- Molded reference point on the lid
- Rack measures 5.125×5.125×1.75 cm
Spécifications
- Color:Green
- Désignation:81-Well Storage Rack