Phosphate Buffered Saline, Electron Microscopy Sciences
Phosphate Buffered Saline, Electron Microscopy Sciences
Phosphate Buffered Saline, 1X, 3.78 l, Bottle
Catalog # 102092-224
Supplier #:  19344-20
Phosphate Buffered Saline, Electron Microscopy Sciences
Catalog # 102092-224
Supplier:  
CAS Number:  
Phosphate Buffered Saline, Electron Microscopy Sciences
Phosphate Buffered Saline, Electron Microscopy Sciences
Phosphate Buffered Saline, 1X, 3.78 l, Bottle
CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
PBS SOLUTION (PH 7.4) 1INX Catalog #101449-092
Product 101449-092 is similar to 102092-224 but manufacturer and specifications may vary.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
PBS is commonly used in biochemistry.
It is a salty solution containing calcium chloride, sodium phosphate and potassium phosphate. PBS is isotonic and non-toxic to cells. 1X PBS final concentration is 8.0 g/l NaCl, 0.2 g/l KCl, 1.42 g/l Na₂HPO₄, 0.24 g/l KH₂PO₄ and pH 7.4.
Specifications
Specifications
  • Type de conditionnement:
    Bottle
  • Désignation:
    Phosphate Buffered Saline, 1X
  • Taille:
    3.78 l