Phosphate Buffered Saline, Electron Microscopy Sciences | Avantor
Phosphate Buffered Saline, Electron Microscopy Sciences
Phosphate Buffered Saline, 1X, 1 l, Bottle
Catalog # 102092-222
Fournisseur #:  19344-10
Phosphate Buffered Saline, Electron Microscopy Sciences
Catalog # 102092-222
Supplier:  
CAS Number:  
Phosphate Buffered Saline, Electron Microscopy Sciences
Phosphate Buffered Saline, Electron Microscopy Sciences
Phosphate Buffered Saline, 1X, 1 l, Bottle
CAS Number:  
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
PBS SOLUTION (PH 7.4) 1INX Catalog #101449-092
Le produit 101449-092 est similaire à 102092-222 mais le fabricant et les spécifications peuvent varier.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
PBS is commonly used in biochemistry.
It is a salty solution containing calcium chloride, sodium phosphate and potassium phosphate. PBS is isotonic and non-toxic to cells. 1X PBS final concentration is 8.0 g/l NaCl, 0.2 g/l KCl, 1.42 g/l Na₂HPO₄, 0.24 g/l KH₂PO₄ and pH 7.4.
Spécifications
Spécifications
  • Type de conditionnement:
    Bottle
  • Désignation:
    Phosphate Buffered Saline, 1X
  • Taille:
    1 l