Phosphate Buffered Saline, Electron Microscopy Sciences | Avantor
Phosphate Buffered Saline, Electron Microscopy Sciences
Phosphate Buffered Saline, 2X, 3.78 l, Bottle
Catalog # 102092-220
Fournisseur #:  19343-40
Phosphate Buffered Saline, Electron Microscopy Sciences
Catalog # 102092-220
Supplier:  
CAS Number:  
Phosphate Buffered Saline, Electron Microscopy Sciences
Phosphate Buffered Saline, Electron Microscopy Sciences
Phosphate Buffered Saline, 2X, 3.78 l, Bottle
CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
PBS is commonly used in biochemistry.
It is a salty solution containing calcium chloride, sodium phosphate and potassium phosphate. PBS is isotonic and non-toxic to cells. 1X PBS final concentration is 8.0 g/l NaCl, 0.2 g/l KCl, 1.42 g/l Na₂HPO₄, 0.24 g/l KH₂PO₄ and pH 7.4.
Spécifications
Spécifications
  • Type de conditionnement:
    Bottle
  • Désignation:
    Phosphate Buffered Saline, 2X
  • Taille:
    3.78 l