Bouin's Solution, Electron Microscopy Sciences
Bouin's fixative is used on biopsy specimens of the gastrointestinal tract.
- Used on biopsy specimens
- Fixation and staining fluid
Bouin's Solution is used as a fixation and staining fluid. In prefilled leak-proof histo-containers it is ready-to-use. Formulated with Picric Acid saturated aqueous solution 75%, Formalin 25% and Glacial Acetic acid 5% in de-ionized water.
Bouin's fixative is excellent for use on biopsy specimens of the gastrointestinal tract. Tissue from the endocrine system are well fixed and many antibodies react well with tissue fixed in Bouin's. Bouin's solution may be used as a routine fixative or a staining solution, but should not be used as a preservative for tissue. Tissue cannot be held in Bouin's solution for extended periods. The maximum fixation time in this solution should be less than 48 hours and should be transferred to alcohol.
- Désignation:Bouin's Solution
- Taille:4 L