3M™ Scotch 810 Magic Tape, Electron Microscopy Sciences | Avantor
3M™ Scotch 810 Magic Tape, Electron Microscopy Sciences
Scotch 810 magic tape, ³/₄"
Catalog # 102091-332
Fournisseur #:  77301
3M™ Scotch 810 Magic Tape, Electron Microscopy Sciences
Catalog # 102091-332
3M™ Scotch 810 Magic Tape, Electron Microscopy Sciences
3M™ Scotch 810 Magic Tape, Electron Microscopy Sciences
Scotch 810 magic tape, ³/₄"
Price
28,62 $
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The ever popular tape that is invisible when applied. Can be written on. Resists yellowing, cracking, and moisture. Ideal for mending, slicing, and many other permanent applications. Tape has 3" core.
Spécifications
  • Désignation:
    Scotch 810 magic tape, ³/₄"