Polyethylene Disposable Gloves, Electron Microscopy Sciences
Product Details & Documents
Low cost, offers good protection to hands from chemical contamination.
- Excellent Sensitivity
- Powderless/sulfur-free
- Ambidextrous
- 1¾ mil thickness
: 100 gloves per roll. Comes in a Dispenser Box.
Spécifications
- Taille du gant:L
- Cat. No.:102091-192
- Disposable / reusable:Disposable
- Ambidextrous/Hand-specific:Ambidextrous