Nunclon™ Multi-Dishes Plates, Electron Microscopy Sciences
Product Details & Documents
Useful in all areas of cell culture including scale-up and cloning.
- Raised well rims to lower risk of cross contamination
- Excellent optical quality
- Nunclon™ Delta certified
- Multidish 6 is available coated with Poly-D-Lysine or Collagen
Spécifications
- Nb de puits:48
- Sterility:Sterile
- Désignation:MultiDish
- Well Shape:Round
- Sterile:Sterile