Nunclon™ Multi-Dishes Plates, Electron Microscopy Sciences
MultiDish, 24, Sterile, Sterile, Round
Catalog # 102091-148
 :  70448
Nunclon™ Multi-Dishes Plates, Electron Microscopy Sciences
Catalog # 102091-148
Nunclon™ Multi-Dishes Plates, Electron Microscopy Sciences
Nunclon™ Multi-Dishes Plates, Electron Microscopy Sciences
MultiDish, 24, Sterile, Sterile, Round
Price
$32.67
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Useful in all areas of cell culture including scale-up and cloning.
  • Raised well rims to lower risk of cross contamination
  • Excellent optical quality
  • Nunclon™ Delta certified
  • Multidish 6 is available coated with Poly-D-Lysine or Collagen
Spécifications
  • Nb de puits:
    24
  • Sterility:
    Sterile
  • Désignation:
    MultiDish
  • Well Shape:
    Round
  • Sterile:
    Sterile