Foil Roll, Electron Microscopy Sciences
Foil Roll 102 mm ( 4") x152.4 m
  102091-128
Foil Roll, Electron Microscopy Sciences
  102091-128
Foil Roll, Electron Microscopy Sciences
Foil Roll, Electron Microscopy Sciences
Foil Roll 102 mm ( 4") x152.4 m
$43.53
Each
Custom-sized pieces of foil can easily be created for covering small surfaces such as beakers, flasks, tubes, and petri dishes.
  • Create custom-size pieces of foil
  • Minimal waste
  • Two sizes available
Designed to work without Mini Bin™ Foil Dispenser, these foil rolls are ideal for autoclaving, freezing, incubating, protecting lab ware, lining, sealing and general storage.
  • Désignation:
    Foil Roll 102 mm ( 4") x152.4 m