Micro Tissue Grinder Kit, Electron Microscopy Sciences
Micro Tissue Grinder Kit, Electron Microscopy Sciences
Complete Kit
  102091-106
Supplier #:  64800
Micro Tissue Grinder Kit, Electron Microscopy Sciences
  102091-106
Micro Tissue Grinder Kit, Electron Microscopy Sciences
Micro Tissue Grinder Kit, Electron Microscopy Sciences
Complete Kit
$2,019.97
Each
This complete selection of Wheaton Micro Tissue Grinders is conveniently packed in a lightweight protective carrying case. The 0.5 ml micro tissue grinder with screw cap can be used for tissue grinding as well as for additional procedures.
  • Complete kit includes 7 micro tissue grinders
  • Made from Wheaton 33 low extractable borosilicate glass
  • Replacement parts available separately
  • Autoclavable
This particular tissue grinder mortar can be safely joined with another threaded glass component if needed, using the Wheaton Connection®. The Kit is packed in high-density polyethylene case with foam inserts.
  • Désignation:
    Complete Kit