AMD Balance Table with Isolation Area, Electron Microscopy Sciences
AMD-SB balance table with isolation area, Stainless steel, 75×75"
  102090-894
 :  6700
AMD Balance Table with Isolation Area, Electron Microscopy Sciences
  102090-894
 :  
AMD Balance Table with Isolation Area, Electron Microscopy Sciences
AMD Balance Table with Isolation Area, Electron Microscopy Sciences
AMD-SB balance table with isolation area, Stainless steel, 75×75"
These workstations remove unwanted vibrations which limit the performance of sensitive instruments such as balances. Instruments are placed directly onto this isolation section of the workstation and isolations are removed by the built-in isolators.
  • Top surface 75x75 cm incoroprates a 30x45 cm vibration isolation platform, which sits flush with the surrounding table surface in a recess
  • Removes up to 99 percent of unwanted vertical and horizontal vibrations
  • Flush working area around balance for supporting hardware
  • Tilting levelling feet on the frame ensure the table sits stabily on the floor
  • 240 satin silicon 0.4Ra ground stainless steel meets international standards for cleanliness
  • Three versions are available, the ST and SS versions can be easily upgraded to the SA at a later date if required
AMD-SA stainless air table features our AMB-30x45 pneumatic platform. It incorporates four low 2 Hz natural frequency rolling diaphragm damped isolators and the platform and table top feature 240 ground satin stainless steel. The isolators are simply inflated via adjustable valves using a bicycle pump.
AMD-SS stainless version features an AMS-30x45 platform with four 7 Hz sorbothane isolators which give an excellent 95 percent isolation at 50Hz. A unique sound deadening core soaks up surface borne vibrations and ensures our tables outerperform simple granite and steel tables by up to 20dB. 240 silicon satin stainless ensures our tables meet all the requirements for use in stringent steel case environments such as cleanrooms and biohazard areas.
AMD-ST and AMD-SB standard table has a white powder epoxy table top surface incorporating a AMT-30 x 40 granite platform featuring four sorbothane isolators. It improves balance performance and is ideal for balances up to 4 to 5 digit The smooth seamless table shape ensures thorough and easy cleaning removing risks such as cross contamination. The stainless steel versions feature 240 satin silicon steel with a very low Ra of 0.4 making them ideal for cleanrooms and as well as being resistant to almost all forms of biological and chemical attack. The frame is produced using tubular ERW steel which combines high rigidity with a closed profile that allows easy and thorough cleaning.
 : Table legs are not included and sold separately. Options, accessories, and components are also available. Contact your Avantor representative for more information.
  • Worksurface Material:
    Stainless steel
  • Désignation:
    AMD-SB balance table with isolation area
  • W×D:
    75×75"