Protector® Evidence Drying Cabinet, Labconco
Protector Evidence Drying Cabinet with Washdown, 5', 230 V (US)
  89260-162
 :   Labconco
 :  3405011
Protector® Evidence Drying Cabinet, Labconco
  89260-162
 :  Labconco
Protector® Evidence Drying Cabinet, Labconco
Protector® Evidence Drying Cabinet, Labconco
Protector Evidence Drying Cabinet with Washdown, 5', 230 V (US)
$22,970.00
Each
The durable, stainless steel-lined Protector® Evidence Drying Cabinet provides a secure ventilated area to dry evidence such as clothing and bedding
Protector Evidence Drying Cabinet with Washdown features a spray nozzle that allows the interior to be washed down for easy clean up and after the cabinet has been properly disinfected. A built-in water pump aids clean up. Note that Evidence Drying Cabinet units with Washdown do not include UV lights.
Protector® Benchtop Evidence Drying Cabinets provide a compact, space-saving method of securely drying evidence. The cabinet's HEPA and carbon filtered exhaust protects personnel from exposure to biohazards while eliminating nuisance odors. The key-activated ultraviolet light provides secondary disinfection of the interior between cases to prevent cross contamination, ensuring integrity of samples for DNA testing.
A variety of accessories for Protector Evidence Drying Cabinets are available, including filters, cleaning supplies, drain kits for washdown units and more. Choose from a variety of accessories for all sizes of Protector Evidence Drying Cabinets. In applicable instances, note the size of the cabinet when ordering prefilters.
The cabinet's HEPA and carbon filtered exhaust protects personnel from exposure to biohazards while eliminating nuisance odors. The key-activated ultraviolet light provides secondary disinfection of the interior between cases to prevent cross contamination, ensuring integrity of samples for DNA testing. The lockable cabinet maintains the chain of custody. Full-length cabinets are available in 3', 4' and 5' widths and benchtop cabinets in 3' width. 3' cabinets have a single door and include 2" diameter locking casters. 4' and 5' cabinets have two doors and include fixed leveling feet.
 : ETL
  • Désignation:
    Protector Evidence Drying Cabinet with Washdown
  • Taille:
    5'
  • Electrical:
    230 V (US)
Accessories
  •  
    Description
  • 97054-324
    Hanging Clips
    Each - $110.00
  •  
    Description
  • 97054-326
    Exhaust Connection Kit
    Each - $721.00
  •  
    Description
  • 97054-328
    Paper liners, tube of 25
    Each - $182.00
  •  
    Description
  • 97054-330
    Drying Cabinet Starter Kit
    Each - $637.00
  •  
    Description
  • 97054-336
    Prefilters, Inlet, Replacement
    Each - $178.00
  •  
    Description
  • 97054-340
    Paper Liners, Tube of 100
    Each - $651.00
  •  
    Description
  • 97054-346
    Security Tags
    Each - $244.00
  •  
    Description
  • 89167-556
    Drain Kit for Washdown Units
    Each - $462.00
  •  
    Description
  • 97054-338
    Prefilters, Exhaust, Replacement
    Each - $210.00
  •  
    Description
  • 89208-232
    Cabinet Prefilter, pkg. 10
    Pack of 10 - $74.70
  •  
    Description
  • 89260-188
    Paper Liners for 5' EDC, pkg. 100
    Pack of 100 - $1,400.00
  •  
    Description
  • 89208-230
    Door Prefilter, pkg. 10
    Pack of 10 - $83.70
  •  
    Description
  • 89260-186
    Floor Paper Liners for 4' EDC, pkg. 100
    Pack of 100 - $1,140.00
  •  
    Description
  • 89260-184
    Floor Paper Liners for 5' EDC, pkg. 25
    Pack of 25 - $413.00
  •  
    Description
  • 89260-182
    Floor Paper Liners for 4' EDC, pkg. 25
    Pack of 25 - $297.00
  •  
    Description
  • 89260-180
    Starter Kit for 5' EDC
    Each - $686.00
  •  
    Description
  • 89260-178
    Starter Kit for 4' EDC
    Each - $644.00
  •  
    Description
  • 89260-176
    Door Prefilter for 5' EDC, pkg. 6
    Pack of 6 - $290.00
  •  
    Description
  • 89260-174
    Door Prefilter for 4' EDC, pkg. 6
    Pack of 6 - $255.00
  •  
    Description
  • 89260-172
    Door Evidence Prefilter for 5' EDC, pkg. 10
    Pack of 10 - $123.00
  •  
    Description
  • 89260-170
    Door Evidence Prefilter for 4' EDC, pkg. 10
    Pack of 10 - $123.00