Protector® Evidence Drying Cabinet, Labconco
Protector® Evidence Drying Cabinet, Labconco
Protector Evidence Drying Cabinet, 4', 115 V
Catalog # 89260-130
Supplier:   Labconco
Supplier #:  3404000
Protector® Evidence Drying Cabinet, Labconco
Catalog # 89260-130
Supplier:  Labconco
Protector® Evidence Drying Cabinet, Labconco
Protector® Evidence Drying Cabinet, Labconco
Protector Evidence Drying Cabinet, 4', 115 V
Price
$16,900.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
The durable, stainless steel-lined Protector® Evidence Drying Cabinet provides a secure ventilated area to dry evidence such as clothing and bedding
Protector Evidence Drying Cabinet with Washdown features a spray nozzle that allows the interior to be washed down for easy clean up and after the cabinet has been properly disinfected. A built-in water pump aids clean up. Note that Evidence Drying Cabinet units with Washdown do not include UV lights.
Protector® Benchtop Evidence Drying Cabinets provide a compact, space-saving method of securely drying evidence. The cabinet's HEPA and carbon filtered exhaust protects personnel from exposure to biohazards while eliminating nuisance odors. The key-activated ultraviolet light provides secondary disinfection of the interior between cases to prevent cross contamination, ensuring integrity of samples for DNA testing.
A variety of accessories for Protector Evidence Drying Cabinets are available, including filters, cleaning supplies, drain kits for washdown units and more. Choose from a variety of accessories for all sizes of Protector Evidence Drying Cabinets. In applicable instances, note the size of the cabinet when ordering prefilters.
The cabinet's HEPA and carbon filtered exhaust protects personnel from exposure to biohazards while eliminating nuisance odors. The key-activated ultraviolet light provides secondary disinfection of the interior between cases to prevent cross contamination, ensuring integrity of samples for DNA testing. The lockable cabinet maintains the chain of custody. Full-length cabinets are available in 3', 4' and 5' widths and benchtop cabinets in 3' width. 3' cabinets have a single door and include 2" diameter locking casters. 4' and 5' cabinets have two doors and include fixed leveling feet.
Certifications: ETL
Specifications
  • Désignation:
    Protector Evidence Drying Cabinet
  • Taille:
    4'
  • Electrical:
    115 V
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Hanging Clips
    Hanging Clips
    97054-324
    Log in for availability
    Each - $110.00
Product Information
  • Catalog #
    Availability
    Price
  • Exhaust Connection Kit
    Exhaust Connection Kit
    97054-326
    Log in for availability
    Each - $721.00
Product Information
  • Catalog #
    Availability
    Price
  • Paper liners, tube of 25
    Paper liners, tube of 25
    97054-328
    Log in for availability
    Each - $182.00
Product Information
  • Catalog #
    Availability
    Price
  • Drying Cabinet Starter Kit
    Drying Cabinet Starter Kit
    97054-330
    Log in for availability
    Each - $637.00
Product Information
  • Catalog #
    Availability
    Price
  • Prefilters, Inlet, Replacement
    Prefilters, Inlet, Replacement
    97054-336
    Log in for availability
    Each - $178.00
Product Information
  • Catalog #
    Availability
    Price
  • Paper Liners, Tube of 100
    Paper Liners, Tube of 100
    97054-340
    Log in for availability
    Each - $651.00
Product Information
  • Catalog #
    Availability
    Price
  • Security Tags
    Security Tags
    97054-346
    Log in for availability
    Each - $244.00
Product Information
  • Catalog #
    Availability
    Price
  • Drain Kit for Washdown Units
    Drain Kit for Washdown Units
    89167-556
    Log in for availability
    Each - $462.00
Product Information
  • Catalog #
    Availability
    Price
  • Prefilters, Exhaust, Replacement
    Prefilters, Exhaust, Replacement
    97054-338
    Log in for availability
    Each - $210.00
Product Information
  • Catalog #
    Availability
    Price
  • Cabinet Prefilter, pkg. 10
    Cabinet Prefilter, pkg. 10
    89208-232
    Log in for availability
    Pack of 10 - $74.70
Product Information
  • Catalog #
    Availability
    Price
  • Paper Liners for 5' EDC, pkg. 100
    Paper Liners for 5' EDC, pkg. 100
    89260-188
    Log in for availability
    Pack of 100 - $1,400.00
Product Information
  • Catalog #
    Availability
    Price
  • Door Prefilter, pkg. 10
    Door Prefilter, pkg. 10
    89208-230
    Log in for availability
    Pack of 10 - $83.70
Product Information
  • Catalog #
    Availability
    Price
  • Floor Paper Liners for 4' EDC, pkg. 100
    Floor Paper Liners for 4' EDC, pkg. 100
    89260-186
    Log in for availability
    Pack of 100 - $1,140.00
Product Information
  • Catalog #
    Availability
    Price
  • Floor Paper Liners for 5' EDC, pkg. 25
    Floor Paper Liners for 5' EDC, pkg. 25
    89260-184
    Log in for availability
    Pack of 25 - $413.00
Product Information
  • Catalog #
    Availability
    Price
  • Floor Paper Liners for 4' EDC, pkg. 25
    Floor Paper Liners for 4' EDC, pkg. 25
    89260-182
    Log in for availability
    Pack of 25 - $297.00
Product Information
  • Catalog #
    Availability
    Price
  • Starter Kit for 5' EDC
    Starter Kit for 5' EDC
    89260-180
    Log in for availability
    Each - $686.00
Product Information
  • Catalog #
    Availability
    Price
  • Starter Kit for 4' EDC
    Starter Kit for 4' EDC
    89260-178
    Log in for availability
    Each - $644.00
Product Information
  • Catalog #
    Availability
    Price
  • Door Prefilter for 5' EDC, pkg. 6
    Door Prefilter for 5' EDC, pkg. 6
    89260-176
    Log in for availability
    Pack of 6 - $290.00
Product Information
  • Catalog #
    Availability
    Price
  • Door Prefilter for 4' EDC, pkg. 6
    Door Prefilter for 4' EDC, pkg. 6
    89260-174
    Log in for availability
    Pack of 6 - $255.00
Product Information
  • Catalog #
    Availability
    Price
  • Door Evidence Prefilter for 5' EDC, pkg. 10
    Door Evidence Prefilter for 5' EDC, pkg. 10
    89260-172
    Log in for availability
    Pack of 10 - $123.00
Product Information
  • Catalog #
    Availability
    Price
  • Door Evidence Prefilter for 4' EDC, pkg. 10
    Door Evidence Prefilter for 4' EDC, pkg. 10
    89260-170
    Log in for availability
    Pack of 10 - $123.00