Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
CELLine™ Bioreactor Flasks, Wheaton | Avantor

CELLine™ Bioreactor Flasks, Wheaton

CELLine 1000AD, Adherent Type, Package of 1, 1000 ml, 15 ml
Catalog # 89237-448
Fournisseur #:  WCL1000AD-1
CELLine™ Bioreactor Flasks, Wheaton
Catalog # 89237-448
CELLine™ Bioreactor Flasks, Wheaton
CELLine™ Bioreactor Flasks, Wheaton

CELLine 1000AD, Adherent Type, Package of 1, 1000 ml, 15 ml

Price
$621.53
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 1
Product Details & Documents

Wheaton manufactures a new line of high density cell culture membrane flasks for antibody and protein production. CELLine works well with secreting cell lines such as Hybridoma, CHO, NS0, SF, HEK, and BHK cells. Conventional in vivo or in vitro cell culture methods can be laborious, low density and require significant purification. CELLine dual compartment flasks separate the cell compartment from the bulk media compartment with a 10 kD MWCO dialysis membrane. This allows for longer run times, high density cultures, and a super concentrated harvest. The CELLine flasks were designed to address three areas of limitation:

Handling Requirements – CELLine flasks reduce the labor requirement by requiring less consumable items and allowing longer run times due to their unique metabolite regulating upper membrane. This membrane allows for bulk media storage during operation to ensure constant and regulated nutrient access for the cells.

Cell Densities – CELLine flasks ensure maximum gas exchange by placing the gas permeable lower membrane directly next to the cells. This allows for optimal oxygen and carbon dioxide transfer for metabolizing cells.

Purification – The upper and lower membranes form an optimized compartment for cell proliferation. This concentrates the antibody and proteins of interest and keeps them away from the bulk medium. CELLine flasks also allow for the reduced use of growth factors and serum.

Handling Requirements – CELLine flasks reduce the labor requirement by requiring less consumable items and allowing longer run times due to their unique metabolite regulating upper membrane

Ordering information: Individually Packaged, Gamma Irradiated, Ready to use.

Spécifications
  • Capacité:
    1000 ml
  • Désignation:
    CELLine 1000AD, Adherent Type, Package of 1
  • Cell Growth Area:
    15 ml
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.