Nunc® Barcode Scanners | Avantor
Nunc® Barcode Scanners
Catalog # CA73521-418
Fournisseur #:  330050
Nunc® Barcode Scanners
Catalog # CA73521-418
Nunc® Barcode Scanners
Nunc® Barcode Scanners
Price
$1,870.47
Log in pour la disponibilité
Case of 1
Product Details & Documents
Nunc bar code scanners with plug and play functionality play a critical role in sample tracking
The scanners automatically switch between code 128, code 39, interleave 2 of 5 and several other barcodes. These scanners are durable low-cost, and easy to use.