Snap Caps, Wheaton
Natural cap, 11 mm, Red PTFE/Silicone
  89235-566
 :  242772
Snap Caps, Wheaton
  89235-566
Snap Caps, Wheaton
Snap Caps, Wheaton
Natural cap, 11 mm, Red PTFE/Silicone
$1,529.50
Case of 1,000
Eliminate the need for crimping and decapping tools.
  • Manufactured from polyethylene
Fits the WHEATON E-Z vials® with snap ring top.
  • Septum:
    Red PTFE/Silicone
  • Désignation:
    Natural cap
  • Taille:
    11 mm